Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_126165.1 | NP_569053.1 | 2 | 205 | ||
GenBank mRNA | AK047821.1 | - | 2 | 217 | ||
AK159117.1 | - | 2 | 231 | |||
AK169949.1 | - | 2 | 193 | |||
BC018368.1 | - | 2 | 205 | 3963907 |