Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005077.1 | - | 4, 5 | 749 | ||
AK039267.1 | - | 3, 4 | 409 | |||
BC004802.1 | - | 5, 6 | 580 | 3589325 | ||
BC008184.1 | - | 5, 6 | 580 | 3589205 | ||
S72537.1 | - | 5, 6 | 604 | |||
X03796.1 | - | 5, 6 | 604 |