Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007443.3 | NP_031469.1 | 1 | 156 | ||
GenBank mRNA | AK004907.1 | - | 1 | 157 | ||
BC021660.1 | - | 1 | 175 | 4193922 | ||
D28812.1 | - | 1 | 157 | |||
X68680.1 | - | 1 | 118 |