Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip4k2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054097.3 | NP_473438.1 | 8 | 1109 | ||
GenBank mRNA | AB054591.1 | - | 8 | 1052 | ||
AK148299.1 | - | 8 | 1069 | |||
AK178521.1 | - | 1 | 42 | |||
BC005539.1 | - | 3 | 323 | |||
BC021383.1 | - | 8 | 1042 | 5042933 |