Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pip4k2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054097.3 | NP_473438.1 | 1 | 219 | ||
GenBank mRNA | AB054591.1 | - | 1 | 162 | ||
AK140079.1 | - | 1 | 183 | |||
AK148299.1 | - | 1 | 179 | |||
AK212271.1 | - | 1 | 51 | |||
BC021383.1 | - | 1 | 152 | 5042933 |