Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110783.3 | NP_001104253.1 | 20 | 2668 | ||
NM_001277280.2 | NP_001264209.1 | 20 | 2668 | |||
NM_001277281.2 | NP_001264210.1 | 20 | 2389 | |||
NM_001277284.2 | NP_001264213.1 | 20 | 2389 | |||
NM_001277286.2 | NP_001264215.1 | 20 | 2654 | |||
NM_001277289.2 | NP_001264218.1 | 20 | 2654 | |||
NM_031158.4 | NP_112435.2 | 20 | 2654 | |||
GenBank mRNA | AK147278.1 | - | 20 | 2623 | ||
AK147483.1 | - | 20 | 2650 | |||
AK147597.1 | - | 20 | 2602 | |||
AK158131.1 | - | 20 | 2661 | |||
BC079910.1 | - | 20 | 2360 | 30664024 | ||
BC138029.1 | - | 20 | 2388 | 8861048 | ||
BC145102.1 | - | 20 | 2388 | |||
BC171944.1 | - | 20 | 2388 | 9054230 | ||
M84756.1 | - | 20 | 2366 | |||
X69063.1 | - | 21 | 2638 | |||
X69065.1 | - | 9 | 1060 |