Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009670.4 | NP_033800.2 | 16 | 2685 | ||
NM_146005.3 | NP_666117.2 | 39 | 5156 | |||
NM_170687.3 | NP_733788.2 | 16 | 2685 | |||
NM_170688.2 | NP_733789.1 | 39 | 5156 | |||
NM_170689.2 | NP_733790.2 | 38 | 5102 | |||
NM_170690.2 | NP_733791.2 | 38 | 5093 | |||
NM_170728.2 | NP_733924.2 | 37 | 5039 | |||
NM_170729.2 | NP_733925.2 | 37 | 5039 | |||
NM_170730.2 | NP_733926.2 | 14 | 2375 | |||
GenBank mRNA | AF212924.1 | - | 14 | 1875 | ||
AK035681.1 | - | 16 | 2413 | |||
AK162007.1 | - | 16 | 2343 | |||
BC021657.1 | - | 37 | 4909 | 4188590 | ||
BC026152.1 | - | 16 | 2287 | |||
BC062122.1 | - | 5 | 2993 | |||
L40631.1 | - | 16 | 2688 | |||
L40632.1 | - | 39 | 4698 | |||
U89274.1 | - | 18 | 2302 | |||
U89275.1 | - | 18 | 2302 |