Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009673.2 | NP_033803.1 | 5, 6 | 651 | ||
GenBank mRNA | AK152185.1 | - | 5, 6 | 486 | ||
AK153388.1 | - | 5, 6 | 432 | |||
AK153476.1 | - | 5, 6 | 415 | |||
D63423.1 | - | 5, 6 | 389 | |||
U29396.1 | - | 5, 6 | 408 |