Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013473.4 | NP_038501.2 | 8 | 654 | ||
GenBank mRNA | AK009085.1 | - | 8 | 616 | ||
AK075870.1 | - | 8 | 588 | |||
BC030407.1 | - | 8 | 667 | 5322310 |