Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081037.2 | NP_001074506.1 | 1 | 168 | ||
NM_001242411.1 | NP_001229340.1 | 1 | 168 | |||
GenBank mRNA | AK138257.1 | - | 1 | 169 | ||
BC120892.1 | - | 1 | 161 | 40061454 | ||
BC120893.1 | - | 1 | 161 | 40061455 |