Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042558.1 | NP_001036023.1 | 22, 23 | 3742 | ||
NM_001282947.1 | NP_001269876.1 | 22, 23 | 3666 | |||
NM_009684.2 | NP_033814.2 | 22, 23 | 3699 | |||
GenBank mRNA | AF064071.1 | - | 22, 23 | 3626 | ||
AK220340.1 | - | 22, 23 | 3738 | |||
BC083191.1 | - | 22, 23 | 3617 | |||
BC131683.1 | - | 22, 23 | 3258 | 40111198 | ||
BC131684.1 | - | 22, 23 | 3258 | 40111199 |