Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apba2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291166.1 | NP_001278095.1 | 11 | 1952 | ||
NM_001291167.1 | NP_001278096.1 | 10 | 1898 | |||
NM_007461.2 | NP_031487.1 | 11 | 1949 | |||
GenBank mRNA | BC057620.1 | - | 11 | 1949 | 6402377 | |
BC060269.1 | - | 8 | 1652 | 6810117 | ||
L34676.1 | - | 8 | 1411 |