Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253885.1 | NP_001240814.1 | 15 | 2412 | ||
NM_001253886.1 | NP_001240815.1 | 14 | 2406 | |||
NM_001253887.1 | NP_001240816.1 | 15 | 1412 | |||
NM_009685.3 | NP_033815.1 | 14 | 2220 | |||
GenBank mRNA | AF206720.1 | - | 14 | 2129 | ||
AK030748.1 | - | 14 | 2103 | |||
AK075730.1 | - | 15 | 1392 | |||
AK083830.1 | - | 15 | 2142 | |||
AK144340.1 | - | 15 | 2413 | |||
AK164140.1 | - | 15 | 2137 | |||
BC022732.1 | - | 8 | 787 | |||
BC036956.1 | - | 14 | 2050 | |||
BC048395.1 | - | 13 | 1558 | 6392016 |