Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253885.1 | NP_001240814.1 | 10 | 1772 | ||
NM_001253886.1 | NP_001240815.1 | 9 | 1766 | |||
NM_001253887.1 | NP_001240816.1 | 10 | 772 | |||
NM_009685.3 | NP_033815.1 | 9 | 1580 | |||
GenBank mRNA | AF206720.1 | - | 9 | 1489 | ||
AK030748.1 | - | 9 | 1463 | |||
AK075730.1 | - | 10 | 752 | |||
AK083830.1 | - | 10 | 1502 | |||
AK144340.1 | - | 10 | 1773 | |||
AK164140.1 | - | 10 | 1497 | |||
BC022732.1 | - | 3 | 147 | |||
BC036956.1 | - | 9 | 1410 | |||
BC048395.1 | - | 8 | 918 | 6392016 |