Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007462.3 | NP_031488.2 | 3 | 501 | ||
GenBank mRNA | AK042021.1 | - | 3 | 502 | ||
AK045053.1 | - | 3 | 472 | |||
AK046280.1 | - | 3 | 492 | |||
AK080907.1 | - | 3 | 501 | |||
AK082724.1 | - | 3 | 247 | |||
BC141141.1 | - | 2 | 214 | 9055753 | ||
M88127.1 | - | 2 | 190 |