Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009700.2 | NP_033830.2 | 5 | 1003 | ||
GenBank mRNA | AF469168.1 | - | 5 | 1077 | ||
AF469169.1 | - | 5 | 1259 | |||
AK039777.1 | - | 5 | 1006 | |||
AK045357.1 | - | 4 | 654 | |||
AY484964.1 | - | 5 | 1009 | |||
AY484965.1 | - | 6 | 1193 | |||
AY484966.1 | - | 6 | 1139 | |||
AY728043.1 | - | 5 | 1023 | |||
AY728044.1 | - | 6 | 1207 | |||
AY728045.1 | - | 5 | 1135 | |||
AY803942.1 | - | 6 | 1114 | |||
AY803943.1 | - | 7 | 1244 | |||
AY803944.1 | - | 8 | 1428 | |||
BC024526.1 | - | 5 | 949 | 4985265 | ||
CT010362.1 | - | 4 | 950 | |||
U33012.1 | - | 4 | 965 | |||
U48397.1 | - | 4 | 1085 | |||
U48398.1 | - | 4 | 956 | |||
U48399.1 | - | 6 | 1067 | |||
U88623.1 | - | 5 | 1004 |