Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Araf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159645.1 | NP_001153117.1 | 4 | 399 | ||
NM_009703.2 | NP_033833.1 | 4 | 399 | |||
GenBank mRNA | AB158252.1 | - | 3 | 324 | ||
AK004741.1 | - | 3 | 1678 | |||
AK010060.1 | - | 2 | 150 | |||
AK020547.1 | - | 4 | 365 | |||
AK034861.1 | - | 4 | 394 | |||
AK039044.1 | - | 4 | 365 | |||
AK085823.1 | - | 4 | 365 | |||
AK133797.1 | - | 3 | 1685 | |||
AK135358.1 | - | 4 | 365 | |||
AK163567.1 | - | 4 | 368 | |||
AK169757.1 | - | 4 | 380 | |||
BC004757.1 | - | 4 | 355 | 3154293 |