Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166031.2 | NP_001159503.2 | 4 | 633 | ||
NM_080452.3 | NP_536700.3 | 4 | 690 | |||
GenBank mRNA | AB055389.1 | - | 4 | 681 | ||
AK051184.1 | - | 4 | 690 | |||
AK077219.1 | - | 5 | 859 | |||
AK154131.1 | - | 4 | 684 | |||
AK156412.1 | - | 3 | 1135 | |||
AK160129.1 | - | 4 | 682 | |||
AK165956.1 | - | 4 | 681 | |||
AK171699.1 | - | 3 | 1153 | |||
AK172120.1 | - | 3 | 1153 | |||
AK172432.1 | - | 3 | 1135 | |||
AK210219.1 | - | 1 | 228 | |||
BC033460.1 | - | 3 | 1139 | |||
BC048377.1 | - | 4 | 1052 | |||
BC060503.1 | - | 4 | 666 | 6306321 |