Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009706.2 | NP_033836.2 | 6 | 4456 | ||
GenBank mRNA | BC009135.1 | - | 2 | 131 | ||
BC150823.1 | - | 6 | 4426 | 9087734 | ||
BU613221.1 | - | 2 | 177 |