Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asgr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007493.3 | NP_031519.1 | 4 | 422 | ||
GenBank mRNA | BC011197.1 | - | 4 | 422 | 4194949 | |
X53042.1 | - | 4 | 298 |