Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301230.1 | NP_001288159.1 | 5 | 679 | ||
NM_001301232.1 | NP_001288161.1 | 4 | 572 | |||
NM_023635.6 | NP_076124.1 | 4 | 530 | |||
GenBank mRNA | AB046693.1 | - | 3 | 328 | ||
AB120511.1 | - | 3 | 306 | |||
AB232622.1 | - | 3 | 306 | |||
AF304376.1 | - | 4 | 485 | |||
AK010373.1 | - | 4 | 448 | |||
AK030529.1 | - | 4 | 477 | |||
BC008173.1 | - | 5 | 636 | 3710213 | ||
BC009656.1 | - | 4 | 483 | 3590089 | ||
CV784419.1 | - | 4 | 441 |