Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025093.2 | NP_001020264.1 | 15 | 1758 | ||
NM_001284369.1 | NP_001271298.1 | 13 | 1598 | |||
NM_001284372.1 | NP_001271301.1 | 13 | 1464 | |||
NM_001284373.1 | NP_001271302.1 | 12 | 1401 | |||
NM_001284374.1 | NP_001271303.1 | 12 | 1483 | |||
NM_001284376.1 | NP_001271305.1 | 15 | 1640 | |||
NM_009715.3 | NP_033845.1 | 14 | 1661 | |||
GenBank mRNA | AF483482.1 | - | 11 | 1335 | ||
AF483483.1 | - | 11 | 1335 | |||
AK046184.1 | - | 4 | 519 | |||
AK051591.1 | - | 14 | 1651 | |||
AK139515.1 | - | 13 | 1514 | |||
AK144977.1 | - | 13 | 1438 | |||
AK145887.1 | - | 13 | 1368 | |||
AY902311.1 | - | 10 | 1221 | |||
BC042210.1 | - | 12 | 1434 | 4018323 | ||
BC079883.1 | - | 14 | 1583 | 6839572 | ||
BC082596.1 | - | 12 | 1347 | 30649207 | ||
BC085137.1 | - | 12 | 1426 | |||
M31629.1 | - | 7 | 813 | |||
M77167.1 | - | 9 | 1107 | |||
S76657.1 | - | 1426 | ||||
S76659.1 | - | 1132 | ||||
U46026.1 | - | 4 | 603 |