Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025093.2 | NP_001020264.1 | 13 | 1469 | ||
NM_001284369.1 | NP_001271298.1 | 11 | 1309 | |||
NM_001284370.1 | NP_001271299.1 | 13 | 1469 | |||
NM_001284371.1 | NP_001271300.1 | 12 | 1406 | |||
NM_001284372.1 | NP_001271301.1 | 11 | 1175 | |||
NM_001284373.1 | NP_001271302.1 | 10 | 1112 | |||
NM_001284374.1 | NP_001271303.1 | 10 | 1194 | |||
NM_001284376.1 | NP_001271305.1 | 13 | 1351 | |||
NM_009715.3 | NP_033845.1 | 12 | 1372 | |||
GenBank mRNA | AF483482.1 | - | 9 | 1046 | ||
AF483483.1 | - | 9 | 1046 | |||
AK035438.1 | - | 13 | 1425 | |||
AK046184.1 | - | 2 | 230 | |||
AK051591.1 | - | 12 | 1362 | |||
AK139515.1 | - | 11 | 1225 | |||
AK144977.1 | - | 11 | 1149 | |||
AK145887.1 | - | 11 | 1079 | |||
AK146839.1 | - | 12 | 1319 | |||
AY902311.1 | - | 8 | 932 | |||
BC042210.1 | - | 10 | 1145 | 4018323 | ||
BC079883.1 | - | 12 | 1294 | 6839572 | ||
BC082596.1 | - | 10 | 1058 | 30649207 | ||
BC085137.1 | - | 10 | 1137 | |||
M31629.1 | - | 5 | 524 | |||
M77167.1 | - | 7 | 818 | |||
S76657.1 | - | 1137 | ||||
S76659.1 | - | 843 | ||||
U46026.1 | - | 2 | 314 |