Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007502.4 | NP_031528.1 | 1 | 195 | ||
GenBank mRNA | AK032516.1 | - | 1 | 212 | ||
AK151700.1 | - | 1 | 212 | |||
AK151841.1 | - | 1 | 211 | |||
AK159532.1 | - | 1 | 212 | |||
AK161282.1 | - | 1 | 227 | |||
AK168273.1 | - | 1 | 211 | |||
AK183986.1 | - | 1 | 85 | |||
AK186227.1 | - | 1 | 87 | |||
AK190411.1 | - | 1 | 87 | |||
BC079916.1 | - | 1 | 200 | 30665138 | ||
U59761.1 | - | 1 | 198 |