Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289445.1 | NP_001276374.1 | 4 | 621 | ||
NM_001289446.1 | NP_001276375.1 | 4 | 351 | |||
NM_015731.3 | NP_056546.2 | 4 | 421 | |||
GenBank mRNA | AF152243.1 | - | 4 | 589 | ||
AK082271.1 | - | 4 | 347 | |||
AK089064.1 | - | 4 | 527 | |||
AK198418.1 | - | 3 | 217 | |||
AK201778.1 | - | 3 | 217 | |||
BC006949.1 | - | 2 | 123 | |||
BC021814.1 | - | 4 | 421 | |||
BC075718.1 | - | 4 | 549 | |||
BC094551.1 | - | 4 | 391 | 6813369 | ||
BY240030.1 | - | 4 | 352 |