Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289445.1 | NP_001276374.1 | 11 | 1253 | ||
NM_001289446.1 | NP_001276375.1 | 11 | 983 | |||
NM_015731.3 | NP_056546.2 | 11 | 1053 | |||
GenBank mRNA | AF152243.1 | - | 11 | 1221 | ||
AK082271.1 | - | 11 | 979 | |||
AK089064.1 | - | 11 | 1159 | |||
BC006949.1 | - | 9 | 755 | |||
BC021814.1 | - | 11 | 1053 | |||
BC075718.1 | - | 11 | 1181 | |||
BC094551.1 | - | 11 | 1023 | 6813369 |