Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cspg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139001.2 | NP_620570.2 | 4 | 4247 | ||
GenBank mRNA | AF352400.1 | - | 4 | 4191 | ||
AK052394.1 | - | 4 | 4319 | |||
AK220559.1 | - | 2 | 1168 |