Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007572.2 | NP_031598.2 | 3 | 573 | ||
GenBank mRNA | AK002655.1 | - | 3 | 499 | ||
AK136737.1 | - | 3 | 499 | |||
AK150771.1 | - | 3 | 499 | |||
AK159210.1 | - | 3 | 499 | |||
AK159343.1 | - | 3 | 494 | |||
BC002086.1 | - | 3 | 532 | 3592169 | ||
BC020096.1 | - | 2 | 495 | |||
X58861.1 | - | 2 | 496 |