Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007573.2 | NP_031599.2 | 5 | 733 | ||
GenBank mRNA | AF300619.1 | - | 5 | 672 | ||
AJ001101.1 | - | 5 | 678 | |||
AK010601.1 | - | 5 | 671 | |||
AK010746.1 | - | 5 | 697 | |||
AK011868.1 | - | 5 | 724 | |||
AK169764.1 | - | 5 | 716 | |||
BC038075.1 | - | 5 | 717 | 3154276 |