Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ciita | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF042158.1 | - | 2 | 274 | ||
AF042159.1 | - | 3 | 580 | |||
AF100709.1 | - | 2 | 457 | |||
AF100710.1 | - | 2 | 173 | |||
AK040723.1 | - | 2 | 336 | |||
AK138822.1 | - | 2 | 366 | |||
AK143956.1 | - | 2 | 445 | |||
AK153609.1 | - | 2 | 270 | |||
AK155184.1 | - | 2 | 297 | |||
AK155311.1 | - | 2 | 366 | |||
BC107330.1 | - | 2 | 215 | |||
BC107331.1 | - | 2 | 215 | |||
U60653.1 | - | 2 | 234 |