Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159533.2 | NP_001153005.1 | 30 | 4418 | ||
NM_001255998.2 | NP_001242927.1 | 31 | 4241 | |||
NM_001255999.2 | NP_001242928.1 | 30 | 4166 | |||
NM_001256000.2 | NP_001242929.1 | 31 | 4493 | |||
NM_001256001.2 | NP_001242930.1 | 31 | 4502 | |||
NM_001290335.1 | NP_001277264.1 | 26 | 3515 | |||
NM_009781.4 | NP_033911.2 | 30 | 4166 | |||
GenBank mRNA | AB259049.1 | - | 30 | 3988 | ||
BC145104.1 | - | 30 | 4112 | 9053652 | ||
FM872410.1 | - | 31 | 4063 | |||
FM872411.1 | - | 30 | 3988 | |||
FM872412.1 | - | 31 | 3973 | |||
FM872413.1 | - | 31 | 3982 | |||
M57973.1 | - | 4 | 415 | |||
U17869.1 | - | 26 | 3515 |