Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110843.1 | NP_001104313.1 | 17, 18 | 1846 | ||
NM_001110844.1 | NP_001104314.1 | 17, 18 | 1846 | |||
NM_001110845.1 | NP_001104315.1 | 17, 18 | 1846 | |||
NM_001110846.1 | NP_001104316.1 | 17, 18 | 1846 | |||
NM_009784.2 | NP_033914.1 | 17, 18 | 1846 | |||
GenBank mRNA | AK014825.1 | - | 12, 13 | 1039 | ||
BC115871.1 | - | 18, 19 | 1621 | 40061614 | ||
U73483.1 | - | 17, 18 | 1753 | |||
U73484.1 | - | 17, 18 | 1753 | |||
U73485.1 | - | 17, 18 | 1753 | |||
U73486.1 | - | 17, 18 | 1753 | |||
U73487.1 | - | 17, 18 | 1753 |