Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252533.1 | NP_001239462.1 | 4 | 482 | ||
NM_023116.4 | NP_075605.1 | 5 | 613 | |||
GenBank mRNA | AK020806.1 | - | 4 | 558 | ||
AK034054.1 | - | 5 | 614 | |||
AK044560.1 | - | 4 | 1132 | |||
AK046048.1 | - | 4 | 470 | |||
AK078220.1 | - | 4 | 338 | |||
AK082315.1 | - | 5 | 497 | |||
AK143332.1 | - | 3 | 289 | |||
AM259383.2 | - | 4 | 482 | |||
BC109156.1 | - | 5 | 545 | 40047524 | ||
BC109157.1 | - | 5 | 545 | 40047531 | ||
FM872406.1 | - | 4 | 438 | |||
FM872407.1 | - | 4 | 544 | |||
FM872408.1 | - | 5 | 612 | |||
KJ789960.1 | - | 4 | 315 | |||
L20343.1 | - | 4 | 392 |