Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252533.1 | NP_001239462.1 | 4, 5 | 585 | ||
NM_023116.4 | NP_075605.1 | 5, 6 | 716 | |||
GenBank mRNA | AK020806.1 | - | 4, 5 | 661 | ||
AK034054.1 | - | 5, 6 | 717 | |||
AK078220.1 | - | 4, 5 | 441 | |||
AM259383.2 | - | 4, 5 | 585 | |||
BC109156.1 | - | 5, 6 | 648 | 40047524 | ||
BC109157.1 | - | 5, 6 | 648 | 40047531 | ||
FM872406.1 | - | 4, 5 | 541 | |||
FM872407.1 | - | 4, 5 | 647 | |||
FM872408.1 | - | 5, 6 | 715 | |||
KJ789960.1 | - | 4, 5 | 418 | |||
L20343.1 | - | 4, 5 | 495 |