Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037099.2 | NP_001032176.1 | 14 | 1688 | ||
NM_001285426.1 | NP_001272355.1 | 14 | 1472 | |||
NM_001285427.1 | NP_001272356.1 | 14 | 1768 | |||
NM_001285428.1 | NP_001272357.1 | 13 | 1397 | |||
NM_146123.3 | NP_666235.1 | 13 | 1608 | |||
GenBank mRNA | AF039417.1 | - | 4 | 465 | ||
AK032311.1 | - | 14 | 1911 | |||
AK038633.1 | - | 14 | 1724 | |||
AK043850.1 | - | 13 | 1551 | |||
AK079616.1 | - | 13 | 1558 | |||
AK147338.1 | - | 14 | 1662 | |||
AK158606.1 | - | 13 | 1399 | |||
BC026479.1 | - | 13 | 1394 | 4501980 |