Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007583.2 | NP_031609.1 | 4 | 1289 | ||
GenBank mRNA | AF077739.1 | - | 4 | 1289 | ||
AK045362.1 | - | 2037 | ||||
BC103563.1 | - | 4 | 970 | 40045028 | ||
BC103564.1 | - | 4 | 969 | 40045029 | ||
BC103664.1 | - | 4 | 969 | 40045030 |