Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301067.1 | NP_001287996.1 | 2 | 315 | ||
NM_007598.4 | NP_031624.2 | 2 | 299 | |||
GenBank mRNA | AK053351.1 | - | 2 | 123 | ||
AK128889.1 | - | 2 | 121 | |||
AK137222.1 | - | 2 | 104 | |||
AK144094.1 | - | 2 | 107 | |||
AK154002.1 | - | 2 | 319 | |||
AK156556.1 | - | 2 | 133 | |||
AK158150.1 | - | 2 | 129 | |||
AK162629.1 | - | 2 | 104 | |||
AK165565.1 | - | 2 | 102 | |||
AK169652.1 | - | 2 | 94 | |||
AK170901.1 | - | 2 | 110 | |||
AK171388.1 | - | 2 | 117 | |||
BC005446.1 | - | 1 | 98 | 3600514 | ||
BC005472.1 | - | 1 | 91 | 3590890 | ||
L12367.1 | - | 2 | 125 |