Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301067.1 | NP_001287996.1 | 8 | 1020 | ||
NM_007598.4 | NP_031624.2 | 8 | 1004 | |||
GenBank mRNA | AK053351.1 | - | 8 | 828 | ||
AK128889.1 | - | 8 | 826 | |||
AK137222.1 | - | 8 | 809 | |||
AK144094.1 | - | 8 | 812 | |||
AK154002.1 | - | 8 | 1024 | |||
AK156556.1 | - | 8 | 838 | |||
AK158150.1 | - | 7 | 1668 | |||
AK165565.1 | - | 8 | 807 | |||
AK169652.1 | - | 8 | 799 | |||
AK170901.1 | - | 8 | 815 | |||
AK171388.1 | - | 8 | 822 | |||
BC005446.1 | - | 7 | 803 | 3600514 | ||
BC005472.1 | - | 7 | 796 | 3590890 | ||
L12367.1 | - | 8 | 830 |