Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cask | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284503.1 | NP_001271432.1 | 2 | 565 | ||
NM_001284504.1 | NP_001271433.1 | 2 | 565 | |||
NM_001284505.1 | NP_001271434.1 | 2 | 565 | |||
NM_009806.3 | NP_033936.2 | 2 | 565 | |||
GenBank mRNA | BC141292.1 | - | 2 | 249 | 9055904 | |
BC145620.1 | - | 2 | 249 | 9054188 | ||
CJ060796.1 | - | 2 | 551 | |||
Y17137.1 | - | 2 | 567 | |||
Y17138.1 | - | 2 | 567 |