Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009809.5 | NP_033939.1 | 6 | 795 | ||
GenBank mRNA | AF092997.1 | - | 5 | 640 | ||
AJ007750.1 | - | 6 | 660 | |||
AK081645.1 | - | 6 | 748 | |||
BC101959.1 | - | 6 | 733 | 40044873 | ||
BC101960.1 | - | 6 | 733 | 40044874 | ||
BC103555.1 | - | 6 | 733 | 40044870 | ||
BC103556.1 | - | 6 | 733 | 40044871 |