Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009818.1 | NP_033948.1 | 4, 5 | 541 | ||
GenBank mRNA | AK004623.1 | - | 4, 5 | 499 | ||
AK036752.1 | - | 4, 5 | 566 | |||
AK148496.1 | - | 4, 5 | 538 | |||
AK149721.1 | - | 4, 5 | 556 | |||
AK149738.1 | - | 4, 5 | 556 | |||
AK159339.1 | - | 4, 5 | 556 | |||
AK168635.1 | - | 4, 5 | 566 | |||
BC048163.1 | - | 4, 5 | 529 | 6418358 | ||
BC060174.1 | - | 4, 5 | 537 | |||
BC068316.1 | - | 4, 5 | 533 | 30106659 | ||
BC105576.1 | - | 4, 5 | 521 | 3967124 | ||
D90362.1 | - | 4, 5 | 532 | |||
X59990.1 | - | 4, 5 | 541 |