Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cav3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007617.3 | NP_031643.1 | 2 | 594 | ||
GenBank mRNA | AA434625.1 | - | 1 | 46 | ||
BC024383.1 | - | 2 | 604 | 4988598 | ||
CJ057698.1 | - | 2 | 371 | |||
U36579.1 | - | 2 | 408 |