Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007634.4 | NP_031660.3 | 5, 6 | 631 | ||
GenBank mRNA | AK030298.1 | - | 570 | |||
AK031613.1 | - | 5, 6 | 571 | |||
AK032961.1 | - | 5, 6 | 557 | |||
AK082485.1 | - | 5, 6 | 599 | |||
AK085183.1 | - | 5, 6 | 568 | |||
AK159594.1 | - | 5, 6 | 632 | |||
AK162447.1 | - | 5, 6 | 571 | |||
AK169263.1 | - | 5, 6 | 582 | |||
BC021658.1 | - | 5, 6 | 551 | |||
BC037662.1 | - | 5, 6 | 583 | 5353169 | ||
U20612.1 | - | 5, 6 | 569 | |||
U20636.1 | - | 5, 6 | 686 | |||
Z47766.1 | - | 5, 6 | 571 |