Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009831.2 | NP_033961.1 | 2 | 408 | ||
GenBank mRNA | AK128957.1 | - | 2 | 396 | ||
AK136645.1 | - | 2 | 146 | |||
AK143114.1 | - | 2 | 399 | |||
AK147026.1 | - | 2 | 396 | |||
AK153392.1 | - | 2 | 340 | |||
AK159429.1 | - | 2 | 338 | |||
AK159831.1 | - | 2 | 343 | |||
AK160411.1 | - | 2 | 383 | |||
AK167052.1 | - | 2 | 340 | |||
AK167486.1 | - | 2 | 335 | |||
AK170397.1 | - | 2 | 396 | |||
BC005534.1 | - | 1 | 58 | |||
BC006609.1 | - | 2 | 292 | |||
L49507.1 | - | 2 | 351 | |||
Z37110.1 | - | 2 | 351 |