Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009831.2 | NP_033961.1 | 6 | 1016 | ||
GenBank mRNA | AK128957.1 | - | 6 | 1004 | ||
AK136645.1 | - | 6 | 753 | |||
AK143114.1 | - | 6 | 1007 | |||
AK147026.1 | - | 6 | 1004 | |||
AK153392.1 | - | 6 | 948 | |||
AK159429.1 | - | 6 | 946 | |||
AK159831.1 | - | 6 | 951 | |||
AK160411.1 | - | 6 | 991 | |||
AK167052.1 | - | 6 | 948 | |||
AK167486.1 | - | 6 | 943 | |||
AK170397.1 | - | 6 | 1008 | |||
BC005534.1 | - | 5 | 666 | |||
BC006609.1 | - | 6 | 899 | |||
L49507.1 | - | 6 | 959 | |||
Z37110.1 | - | 6 | 959 |