Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017367.3 | NP_059063.2 | 7 | 1434 | ||
GenBank mRNA | AF005886.2 | - | 7 | 1086 | ||
AK050465.1 | - | 7 | 1435 | |||
AK079476.1 | - | 7 | 1375 | |||
AK152614.1 | - | 7 | 1390 | |||
AK177017.1 | - | 2 | 336 | |||
AK178822.1 | - | 2 | 317 | |||
AK181042.1 | - | 2 | 320 | |||
AK184350.1 | - | 2 | 336 | |||
AK188314.1 | - | 2 | 307 | |||
AK208801.1 | - | 2 | 255 | |||
AK214983.1 | - | 2 | 295 | |||
AK219418.1 | - | 2 | 336 | |||
BC003290.1 | - | 7 | 967 | 3583418 | ||
BC029328.1 | - | 7 | 967 |