Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009837.1 | NP_033967.1 | 8 | 996 | ||
GenBank mRNA | AK010634.1 | - | 8 | 964 | ||
AK146242.1 | - | 8 | 966 | |||
AK150601.1 | - | 8 | 943 | |||
AK152173.1 | - | 8 | 943 | |||
AK152665.1 | - | 8 | 964 | |||
AK163369.1 | - | 8 | 951 | |||
AK165229.1 | - | 8 | 964 | |||
AK167026.1 | - | 8 | 964 | |||
AK167262.1 | - | 8 | 933 | |||
AK167657.1 | - | 8 | 941 | |||
AK167846.1 | - | 8 | 941 | |||
AK169339.1 | - | 8 | 943 | |||
AK213770.1 | - | 3 | 269 | |||
AK219702.1 | - | 3 | 249 | |||
BC054773.1 | - | 8 | 956 | 6807250 | ||
L25913.1 | - | 8 | 835 | |||
Z31554.1 | - | 8 | 996 |