Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009837.1 | NP_033967.1 | 5 | 668 | ||
GenBank mRNA | AK010634.1 | - | 5 | 636 | ||
AK146242.1 | - | 5 | 638 | |||
AK150601.1 | - | 5 | 615 | |||
AK152173.1 | - | 5 | 615 | |||
AK152665.1 | - | 5 | 636 | |||
AK163369.1 | - | 5 | 623 | |||
AK165229.1 | - | 5 | 636 | |||
AK167026.1 | - | 5 | 636 | |||
AK167262.1 | - | 5 | 605 | |||
AK167657.1 | - | 5 | 613 | |||
AK167846.1 | - | 5 | 613 | |||
AK169339.1 | - | 5 | 615 | |||
AK219961.1 | - | 2 | 188 | |||
BC054773.1 | - | 5 | 628 | 6807250 | ||
L25913.1 | - | 5 | 507 | |||
Z31554.1 | - | 5 | 668 |