Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cct5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007637.2 | NP_031663.1 | 5 | 711 | ||
GenBank mRNA | AK077753.1 | - | 5 | 669 | ||
AK088184.1 | - | 5 | 645 | |||
AK129056.1 | - | 5 | 617 | |||
AK133857.1 | - | 5 | 669 | |||
AK136493.1 | - | 5 | 663 | |||
AK145445.1 | - | 5 | 680 | |||
AK146405.1 | - | 5 | 683 | |||
AK146425.1 | - | 5 | 663 | |||
AK150390.1 | - | 5 | 630 | |||
AK153911.1 | - | 5 | 687 | |||
AK167895.1 | - | 5 | 652 | |||
AK206127.1 | - | 2 | 129 | |||
BC092233.1 | - | 5 | 681 | |||
BC094427.1 | - | 5 | 658 | 30537966 | ||
Z31555.1 | - | 5 | 711 |