Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001043317.2 | NP_001036782.1 | 6 | 873 | ||
NM_009845.3 | NP_033975.3 | 4 | 748 | |||
GenBank mRNA | AF102134.1 | - | 3 | 517 | ||
AF115400.1 | - | 3 | 517 | |||
AF115401.1 | - | 3 | 517 | |||
AJ250676.1 | - | 3 | 660 | |||
AJ250677.1 | - | 3 | 647 | |||
AJ250678.1 | - | 3 | 629 | |||
AJ250679.1 | - | 3 | 592 | |||
AJ250680.1 | - | 4 | 561 | |||
AJ250681.1 | - | 492 | ||||
AJ250682.1 | - | 4 | 490 | |||
AJ250683.1 | - | 443 | ||||
AJ250684.1 | - | 4 | 406 | |||
AK041230.1 | - | 6 | 882 | |||
AK143835.1 | - | 6 | 811 | |||
AK156735.1 | - | 4 | 747 | |||
AK171589.1 | - | 3 | 554 | |||
AK171990.1 | - | 3 | 868 | |||
AK172301.1 | - | 4 | 704 | |||
BC046957.1 | - | 6 | 799 | |||
BC150807.1 | - | 5 | 832 | 9087718 | ||
L02844.1 | - | 5 | 1019 | |||
L16928.1 | - | 5 | 996 |